Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramine T Sodium Salt Trihydrate Crystalline MP Biomedicals

Catalog No. ICN15011583 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15011583 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15011583 Supplier MP Biomedicals, Inc Supplier No. 0215011583

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 127-65-1
Molecular Formula C7H7ClNNaO2S
Molecular Weight (g/mol) 227.638
InChI Key VDQQXEISLMTGAB-UHFFFAOYSA-N
Synonym N-Chloro-p-toluenesulfonamide
PubChem CID 3641960
ChEBI CHEBI:53767
IUPAC Name sodium;chloro-(4-methylphenyl)sulfonylazanide
SMILES CC1=CC=C(C=C1)S(=O)(=O)[N-]Cl.[Na+]

Specifications

Type Chloramine T Sodium Salt
Physical Form Crystalline
Quantity 250 g
Formula Weight 281.7
Chemical Name or Material Chloramine T Sodium Salt, Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.