Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloramine T Trihydrate

Catalog No. 31224250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-224-250G 250 g
31-224-100G 100 g
31-224-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-224-250G Supplier Sigma Aldrich Supplier No. 31224250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloramine T Trihydrate

Specifications

CAS 7080-50-4
Melting Point 167°C to 170°C
Molecular Formula C7H7ClNNaO2S -+ 3H2O
Linear Formula CH3C6H4SO2NClNa -+ 3H2O
MDL Number MFCD00149066
Quantity 250 g
Synonym N-Chloro-p-toluenesulfonamide sodium salt
Molecular Weight (g/mol) 281.69 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.