Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramine T Trihydrate, Fisher Chemical

Catalog No. o1779250 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O1779-250 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. O1779-250
This item is not returnable. View return policy

Chemical Identifiers

CAS 7080-50-4
Molecular Formula C7H13ClNNaO5S
Molecular Weight (g/mol) 281.68
MDL Number MFCD00149066
InChI Key RNROOTWAJBXJBD-UHFFFAOYSA-M
PubChem CID 122197679
IUPAC Name sodium (chloroimino)(4-methylphenyl)-λ⁶-sulfanoylolate trihydrate
SMILES O.O.O.[Na+].CC1=CC=C(C=C1)S([O-])(=O)=NCl

Specifications

Type Chloramine T Trihydrate
Physical Form Powder/Solid
pH 8 to 10
Melting Point 167°C to 170°C
Packaging Glass Bottle
Color White
Quantity 250 g
Formula Weight 281.69
Chemical Name or Material Chloramine T Trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.