Learn More
Alkali Scientific Chloramphenicol, 100 G
SDP
Catalog No. 5000660267
Encompass_Preferred
Chloramphenicol is a broad-spectrum antibiotic supplied in a 100 gram laboratory-grade format for microbiological and biochemical applications. It functions by inhibiting bacterial protein synthesis through binding to the 50S ribosomal subunit, preventing peptide bond formation. This mechanism makes it effective against a wide range of Gram-positive and Gram-negative bacteria. Chloramphenicol is commonly used in selective culture media, molecular biology applications, and antimicrobial research where bacterial growth suppression is required. Due to its stability and broad activity, it is frequently utilized in laboratory workflows involving cloning, plasmid selection, and microbiological testing procedures.
Catalog No. 50-006-60267 Supplier Alkali Scientific Supplier No. CHL001CP
Chloramphenicol is a broad-spectrum antibiotic supplied in a 100 gram laboratory-grade format for microbiological and biochemical applications. It functions by inhibiting bacterial protein synthesis through binding to the 50S ribosomal subunit, preventing peptide bond formation. This mechanism makes it effective against a wide range of Gram-positive and Gram-negative bacteria. Chloramphenicol is commonly used in selective culture media, molecular biology applications, and antimicrobial research where bacterial growth suppression is required. Due to its stability and broad activity, it is frequently utilized in laboratory workflows involving cloning, plasmid selection, and microbiological testing procedures.