Learn More
Selleck Chemical LLC Chloramphenicol S1677-1g
SDP
Catalog No. 503109999
Encompass_Preferred
Chloramphenicol (Chloromycetin) is a bacteriostatic by inhibiting protein synthesis
Catalog No. 50-310-9999 Supplier Selleck Chemical LLC Supplier No. S16771G
Chloramphenicol (Chloromycetin) is a bacteriostatic by inhibiting protein synthesis