Learn More
Apexbio Technology LLC Chloramphenicol(Synonyms: Chloromycetin, Levomycetin, Chlornitromycin, Fenicol, Amphicol, Kemicetine), 10mM (in 1mL DMSO), CAS: 56-75-7.
SDP
Catalog No. 5000060820
Encompass_Preferred
Chloramphenicol (CAS 56-75-7) is a small molecule antibiotic commonly utilized in molecular biology research It exerts anti-microbial activity by specifically binding to the bacterial 50S ribosomal subunit thereby inhibiting the peptidyl transferase reaction and subsequently blocking protein synthesis At elevated concentrations chloramphenicol may also impede DNA synthesis in eukaryotic cells In plasmid selection assays typical effective concentrations are approximately 25 g/ml for stringent plasmids and 170 g/ml for relaxed plasmids For laboratory storage chloramphenicol solutions are maintained at 4 C
Catalog No. 50-000-60820 Supplier Apexbio Technology LLC Supplier No. A25125.1
Chloramphenicol (CAS 56-75-7) is a small molecule antibiotic commonly utilized in molecular biology research It exerts anti-microbial activity by specifically binding to the bacterial 50S ribosomal subunit thereby inhibiting the peptidyl transferase reaction and subsequently blocking protein synthesis At elevated concentrations chloramphenicol may also impede DNA synthesis in eukaryotic cells In plasmid selection assays typical effective concentrations are approximately 25 g/ml for stringent plasmids and 170 g/ml for relaxed plasmids For laboratory storage chloramphenicol solutions are maintained at 4 C