Learn More
Zymo Research Corporation Chloramphenicol (25 ml) 10mg/ml in Ethanol (5 x 5 ml)
SDP
Supplier:  Zymo Research Corporation A100225
Encompass
Chloramphenicol is effective against both Gram (+) and (-) bacteria and some mycobacteria. Resistance to chloramphenicol is conferred by the cat gene which encodes an acetyltransferase that acetylates and inactivates the antibiotic. Chloramphenicol is commonly used for the amplification of vectors in Gram (-) bacteria.
Catalog No. 50-195-7626