Learn More
IBI SCIENTIFIC 3P Chloramphenicol Antibiotic | 56-75-7 | MFCD00078159 | 25 g
Catalog No. NC0753446
Encompass_Preferred
Chloramphenicol - 25gm Chloramphenicol is used in the amplification of plasmids and colony hybridizations. Antibiotics dissolved in ethanol need not be sterilized.
Catalog No. NC0753446 Supplier IBI SCIENTIFIC 3P Supplier No. IB02080
Chloramphenicol - 25gm Chloramphenicol is used in the amplification of plasmids and colony hybridizations. Antibiotics dissolved in ethanol need not be sterilized.