Learn More
Medchemexpress LLC Chloramphenicol 3-acetate | 10318-16-8 | 365.17 | 1 MG
SDP
Catalog No. 5000435995
Encompass_Preferred
Chloramphenicol 3-acetate is the main intermediate in the biodegradation of CAP, formed by the acetylation of the 3-hydroxy group of CAP through chloramphenicol acetyltransferase. This compound represents a common resistance mechanism that microbes employ against chloramphenicol.
  • Main intermediate in the biodegradation of CAP.
  • Formed via acetylation of CAP's 3-hydroxy group by chloramphenicol acetyltransferase.
  • Functions as a common microbial resistance mechanism against chloramphenicol.
Catalog No. 50-004-35995 Supplier Medchemexpress LLC Supplier No. HY1138971MG
Chloramphenicol 3-acetate is the main intermediate in the biodegradation of CAP, formed by the acetylation of the 3-hydroxy group of CAP through chloramphenicol acetyltransferase. This compound represents a common resistance mechanism that microbes employ against chloramphenicol.
  • Main intermediate in the biodegradation of CAP.
  • Formed via acetylation of CAP's 3-hydroxy group by chloramphenicol acetyltransferase.
  • Functions as a common microbial resistance mechanism against chloramphenicol.