Learn More
Medchemexpress LLC Chloramphenicol 3-acetate | 10318-16-8 | MFCD28898459 | >95.0% | 365.17 | C13H14Cl2N2O6 | 5 MG
SDP
Catalog No. 5000264822
Encompass_Preferred
Chloramphenicol 3-acetate (CAS 10318-16-8) is the 3-O-acetylated derivative of chloramphenicol used as an intermediate and reference reagent in biochemical and bacterial metabolism studies. Supplied in small research quantities; purity and storage recommendations are detailed in the certificate of analysis.
  • Acetylated derivative of chloramphenicol.
  • Commonly used as an intermediate in biodegradation and metabolism studies.
  • Provided in small research quantities (for example, 1 mg and 5 mg).
  • Chemical formula C13H14Cl2N2O6; molecular weight 365.17.
  • Certificate of analysis available for purity and storage information.
Catalog No. 50-002-64822 Supplier Medchemexpress LLC Supplier No. HY1138975MG
Chloramphenicol 3-acetate (CAS 10318-16-8) is the 3-O-acetylated derivative of chloramphenicol used as an intermediate and reference reagent in biochemical and bacterial metabolism studies. Supplied in small research quantities; purity and storage recommendations are detailed in the certificate of analysis.
  • Acetylated derivative of chloramphenicol.
  • Commonly used as an intermediate in biodegradation and metabolism studies.
  • Provided in small research quantities (for example, 1 mg and 5 mg).
  • Chemical formula C13H14Cl2N2O6; molecular weight 365.17.
  • Certificate of analysis available for purity and storage information.