Learn More
Crescent Chemical Co Inc CHLORAMPHENICOL
SDP
Catalog No. 50247422
Encompass_Preferred
Chloramphenicol research grade, Ph. Eur. Blocks bacterial protein synthesis by inhibiting the peptidyl transferase activity of the 50S ribosomal subunit. Determination in serum by HPLC (1,2), in meat (3). Molecular basis for chloramphenicol resistance (5,6), effec
Catalog No. 50-247-422 Supplier Crescent Chemical Co Inc Supplier No. SE16785.02
Chloramphenicol research grade, Ph. Eur. Blocks bacterial protein synthesis by inhibiting the peptidyl transferase activity of the 50S ribosomal subunit. Determination in serum by HPLC (1,2), in meat (3). Molecular basis for chloramphenicol resistance (5,6), effec