Learn More
Crescent Chemical Co Inc CHLORAMPHENICOL
SDP
Supplier:  Crescent Chemical Co Inc SE16785.03
Encompass_Preferred
Chloramphenicol research grade, Ph. Eur. Blocks bacterial protein synthesis by inhibiting the peptidyl transferase activity of the 50S ribosomal subunit. Determination in serum by HPLC (1,2), in meat (3). Molecular basis for chloramphenicol resistance (5,6), effec
Catalog No. 50-247-423