Learn More
Zymo Research Corporation Chloramphenicol Solution, 1x5 ml
SDP
Catalog No. 501957627
Encompass
Chloramphenicol is effective against both Gram (+) and (-) bacteria and some mycobacteria. Resistance to chloramphenicol is conferred by the cat gene which encodes an acetyltransferase that acetylates and inactivates the antibiotic. Chloramphenicol is commonly used for the amplification of vectors in Gram (-) bacteria.
Catalog No. 50-195-7627 Supplier Zymo Research Corporation Supplier No. A10025
Chloramphenicol is effective against both Gram (+) and (-) bacteria and some mycobacteria. Resistance to chloramphenicol is conferred by the cat gene which encodes an acetyltransferase that acetylates and inactivates the antibiotic. Chloramphenicol is commonly used for the amplification of vectors in Gram (-) bacteria.