Learn More
Medchemexpress LLC Chloramphenicol | 56-75-7 | 100.0% | 250 MG
SDP
Catalog No. 5000405577
Encompass_Preferred
Chloramphenicol is an orally active, potent, and broad-spectrum antibiotic with antibacterial activity. It represses the oxygen-labile transcription factor and hypoxia inducible factor-1 alpha (HIF-1α) in hypoxic A549 and H1299 cells. It also suppresses the mRNA levels of vascular endothelial growth factor (VEGF) and glucose transporter 1, leading to decreased VEGF release. Chloramphenicol can be used for anaerobic infections and lung cancer research.
  • Inhibits HIF-1α pathway in NSCLC cells
  • Induces autophagy in NSCLC cells
  • Inhibits bacterial and mitochondrial protein synthesis
  • Induces matrix metalloproteinase (MMP)-13 expression
  • Activates JNK and PI-3K/Akt signaling
  • Inhibits peptide bond formation by acting on the 50S subunit of bacterial 70S ribosomes
Catalog No. 50-004-05577 Supplier Medchemexpress LLC Supplier No. 250MGHYB0239R
Chloramphenicol is an orally active, potent, and broad-spectrum antibiotic with antibacterial activity. It represses the oxygen-labile transcription factor and hypoxia inducible factor-1 alpha (HIF-1α) in hypoxic A549 and H1299 cells. It also suppresses the mRNA levels of vascular endothelial growth factor (VEGF) and glucose transporter 1, leading to decreased VEGF release. Chloramphenicol can be used for anaerobic infections and lung cancer research.
  • Inhibits HIF-1α pathway in NSCLC cells
  • Induces autophagy in NSCLC cells
  • Inhibits bacterial and mitochondrial protein synthesis
  • Induces matrix metalloproteinase (MMP)-13 expression
  • Activates JNK and PI-3K/Akt signaling
  • Inhibits peptide bond formation by acting on the 50S subunit of bacterial 70S ribosomes