Learn More
Cayman Chemical Chloramphenicol Palmitate, Purity: ≥99%, CAS: 530-43-8, 25 mg
SDP
Catalog No. 502935145
Encompass_Preferred
An orally bioavailable ester prodrug form of the antibiotic chloramphenicol; is hydrolyzed in the small intestine to release chloramphenicol
Catalog No. 50-293-5145 Supplier Cayman Chemical Supplier No. 2666325 MG
An orally bioavailable ester prodrug form of the antibiotic chloramphenicol; is hydrolyzed in the small intestine to release chloramphenicol