Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramphenicol Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111014652 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4652 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4652 Supplier MilliporeSigma Supelco Supplier No. PHR14121G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chloramphenicol is a synthetic antibiotic, isolated from strains of Streptomyces venezuelae. It is widely used against gram-positive and gram-negative bacteria, and is used mainly for ophthalmic and veterinary purposes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis. Stock solutions can be prepared directly in the vial at any recommended concentration. A solution at 50 mg/mL in ethanol yields a clear, very faint, yellow solution. Degradation of chloramphenicol in aqueous solution is catalyzed by general acids and bases. This rate of degradation is independent of the ionic strength and pH.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Neat
Synonym Chloramphenicol; D-(-)-threo-2,2-Dichloro-N-[beta-hydroxy-alpha-(hydroxymethyl)-beta-(4-nitrophenyl)ethyl]acetamide; D-(-)-threo-2-Dichloroacetamido-1-(4-nitrophenyl)-1,3-propanediol; D-threo-2,2-Dichloro-N-[beta-hydroxy-alpha-(hydroxymethyl)-4-nitrophenethyl]acetamide
Quantity 1 g
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.