Learn More
Teknova 25mg/mL Chloramphenicol in Methanol. 4mL, 25-Pack, Sterile. 4 Pack.
SDP
Catalog No. 502387477
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-238-7477 Supplier Teknova Supplier No. C03164PK
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.