Learn More
Medchemexpress LLC Chloramphenicol (Standard) | 56-75-7 | 99.7% | 25 MG
SDP
Catalog No. 5000450994
Encompass_Preferred
Chloramphenicol is an analytical standard intended for research and analytical applications. It is an orally active, potent, and broad-spectrum antibiotic with antibacterial activity. It represses the oxygen-labile transcription factor and hypoxia inducible factor-1 alpha (HIF-1α) in hypoxic A549 and H1299 cells, suppressing mRNA levels of vascular endothelial growth factor (VEGF) and glucose transporter 1, leading to decreased VEGF release.
  • Used as an analytical standard for research and analytical applications.
  • Potent, orally active, broad-spectrum antibiotic.
  • Exhibits antibacterial activity.
  • Represses HIF-1α in hypoxic A549 and H1299 cells.
  • Suppresses mRNA levels of VEGF and glucose transporter 1.
  • Decreases VEGF release.
  • Suitable for anaerobic infections and lung cancer research.
  • Commonly used in HPLC, GC, and MS as a reference standard.
Catalog No. 50-004-50994 Supplier Medchemexpress LLC Supplier No. 25MGHYB0239R
Chloramphenicol is an analytical standard intended for research and analytical applications. It is an orally active, potent, and broad-spectrum antibiotic with antibacterial activity. It represses the oxygen-labile transcription factor and hypoxia inducible factor-1 alpha (HIF-1α) in hypoxic A549 and H1299 cells, suppressing mRNA levels of vascular endothelial growth factor (VEGF) and glucose transporter 1, leading to decreased VEGF release.
  • Used as an analytical standard for research and analytical applications.
  • Potent, orally active, broad-spectrum antibiotic.
  • Exhibits antibacterial activity.
  • Represses HIF-1α in hypoxic A549 and H1299 cells.
  • Suppresses mRNA levels of VEGF and glucose transporter 1.
  • Decreases VEGF release.
  • Suitable for anaerobic infections and lung cancer research.
  • Commonly used in HPLC, GC, and MS as a reference standard.