Learn More
Cayman Chemical ChloramphenIcol SuccInate 25mg
SDP
Catalog No. 502936571
Encompass_Preferred
A prodrug form of chloramphenicol; a substrate for SDH; is oxidized by human liver and rat liver and kidney mitochondria to release chloramphenicol in vitro; reduces human leukocyte migration in vitro; reduces E. coli growth in rabbit and rat models of pyelonephritis at 150 and 200 mg/kg, respectively; reduces infarct size in a porcine model of myocardial ischemia-reperfusion injury at 20 mg/kg
Catalog No. 50-293-6571 Supplier Cayman Chemical Supplier No. 2545325 MG
A prodrug form of chloramphenicol; a substrate for SDH; is oxidized by human liver and rat liver and kidney mitochondria to release chloramphenicol in vitro; reduces human leukocyte migration in vitro; reduces E. coli growth in rabbit and rat models of pyelonephritis at 150 and 200 mg/kg, respectively; reduces infarct size in a porcine model of myocardial ischemia-reperfusion injury at 20 mg/kg