Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chloramphenicol succinate sodium | 982-57-0 | 1 ML
SDP

Catalog No. 5000417598
Encompass_Preferred

Chloramphenicol succinate sodium is a prodrug of Chloramphenicol, acting as a P2Y14R inhibitor with an IC50 of 1.585 nM. It also serves as a competitive substrate and inhibitor of succinate dehydrogenase (SDH). This compound has shown significant inhibitory effects on colitis and is used in research related to myelosuppression, gray baby syndrome, aplastic anemia, bacterial meningitis, and inflammatory bowel disease. It is slowly oxidized to Chloramphenicol by human bone marrow monocytes and mitochondria from liver and kidney.

  • Molecular weight: 445.18
  • Formula: C15H15Cl2N2NaO8
  • Appearance: Solid, white to light yellow
  • Purity: 97.17% (as per data sheet)
  • Soluble in DMSO (125 mg/mL) and H2O (50 mg/mL)
  • Alleviates DSS-induced colitis in mice
  • Enhances gut barrier integrity via increased tight junction protein expression

Catalog No. 50-004-17598 Supplier Medchemexpress LLC Supplier No. HYN7114A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chloramphenicol succinate sodium is a prodrug of Chloramphenicol, acting as a P2Y14R inhibitor with an IC50 of 1.585 nM. It also serves as a competitive substrate and inhibitor of succinate dehydrogenase (SDH). This compound has shown significant inhibitory effects on colitis and is used in research related to myelosuppression, gray baby syndrome, aplastic anemia, bacterial meningitis, and inflammatory bowel disease. It is slowly oxidized to Chloramphenicol by human bone marrow monocytes and mitochondria from liver and kidney.

  • Molecular weight: 445.18
  • Formula: C15H15Cl2N2NaO8
  • Appearance: Solid, white to light yellow
  • Purity: 97.17% (as per data sheet)
  • Soluble in DMSO (125 mg/mL) and H2O (50 mg/mL)
  • Alleviates DSS-induced colitis in mice
  • Enhances gut barrier integrity via increased tight junction protein expression

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.