Learn More
Gold Biotechnology Inc Chloramphenicol USP Grade
SDP
Catalog No. 5000562110
Encompass_Preferred
Chloramphenicol also known as chlornitromycin is a broad-spectrum bacteriostatic antibiotic derived from Streptomyces venezuelae It is effective against several gram-positive and gram-negative bacteria and commonly used in researching protein synthesis and to select for chloramphenicol-resistant transformed cells or the bacterial CAT /em gene Chloramphenicol functions by binding the 50s ribosomal subunit and inhibiting synthesis of proteins by blocking aminoacyl-tRNA from attaching to the A site
Catalog No. 50-005-62110 Supplier Gold Biotechnology Inc Supplier No. C10525
Chloramphenicol also known as chlornitromycin is a broad-spectrum bacteriostatic antibiotic derived from Streptomyces venezuelae It is effective against several gram-positive and gram-negative bacteria and commonly used in researching protein synthesis and to select for chloramphenicol-resistant transformed cells or the bacterial CAT /em gene Chloramphenicol functions by binding the 50s ribosomal subunit and inhibiting synthesis of proteins by blocking aminoacyl-tRNA from attaching to the A site