Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramphenicol USP Grade, G-Biosciences
SCI_ED

Catalog No. S139773
Change view
Click to view available options
Quantity:
500 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139773 500 g
S157237 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139773 Supplier G-Biosciences Supplier No. RC854
Add to Cart
Add to Cart

Bacteriostatic agent against both gram negative and gram positive bacteria.

Within the cell it binds to the 50S ribosomal subunit and inhibits bacterial protein synthesis by preventing attachment of aminoacyl transfer RNA to its acceptor site on the ribosome, thus preventing peptide bond formation by peptidyl transferase.

Chemical Identifiers

CAS 56-75-7

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 500 g
Percent Purity 98%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.