Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramphenicol USP , Medisca™
GREENER_CHOICESDP

Catalog No. 18048066 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-066 5 kg
18-048-067 500 g
18-048-085 25 g
18-048-086 100 g
18-048-087 25 kg
18-048-107 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-048-066 Supplier Medisca Supplier No. 020302
Add to Cart
Add to Cart

Chloramphenicol USP comes as fine, white to grayish-white or yellowish-white, needle-like crystals or elongated plates.

Chemical Identifiers

CAS 56-75-7

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystals
Color Beige or Gray/Yellow to White
Quantity 5 kg
Percent Purity 97.0 to 103.0%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.