Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloranilic acid

Catalog No. C813625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C813625G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C813625G Supplier Sigma Aldrich Supplier No. C813625G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloranilic acid

Specifications

CAS 87-88-7
Melting Point ≥300°C (lit.)
Molecular Formula C6H2Cl2O4
Linear Formula C6H2Cl2O4
MDL Number MFCD00001596
Quantity 250 mg
Synonym 2,5-Dichloro-3,6-dihydroxy-2,5-cyclohexadiene-1,4-dione; 2,5-Dichloro-3,6-dihydroxy-p-benzoquinone; NSC 6108; NSC 97383
Molecular Weight (g/mol) 208.98
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.