Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD ChloraPrep™ Hi-Lite Orange 3 mL Applicator with Sterile Solution

Catalog No. B930415 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B930415 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B930415 Supplier BD Supplier No. 930415
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easily apply unique formulation for skin preparation

  • Available in 1 mL, Frepp™ 1.5 mL, 3 mL, 10.5 mL
  • 4 sizes and 2 tints
    • Most comprehensive skin antiseptic portfolio
  • Tailor choice to surface area and type of site, color of patient's skin tone
  • Proprietary one-step applicator design can help standardize practice across care settings and reduce variability
  • Evidence of 50+ clinical studies supporting efficacy and safety
TRUSTED_SUSTAINABILITY

Specifications

Product Type Skin Prep
Color Orange
Dimensions (L x W) 4 x 5 in.
Quantity 100/Cs.
Storage Requirements 15°C to 30°C (59°F to 86°F). Product efficacy may degrade outside of this range.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.