Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorate Standard, Ultra Scientific

Catalog No. USICC011
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICC011 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICC011 Supplier Agilent Technologies Supplier No. ICC011
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Density 1
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material Chlorate Standard, 1 analyte at 1000μg/mL in water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.