Learn More
SPEX CERTIPREP LLC 3P CHLORATE STD 100UG/100ML
Catalog No. NC9298819
Encompass_Preferred
(HPS by Zeptometrix) Chlorate standard, Source: NaClO3, Concentration: 100ug/mL, 100mL
Catalog No. NC9298819 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICCLO3100
(HPS by Zeptometrix) Chlorate standard, Source: NaClO3, Concentration: 100ug/mL, 100mL