Learn More
Cayman Chemical Chlorazanil (hydrochloride), Purity: ≥98%, CAS: 2019-25-2, 10 mg
SDP
Catalog No. 502919372
Encompass_Preferred
An analytical reference standard categorized as a diuretic; has been found as a potential doping agent in urine samples from athletes but can be formed from proguanil; intended for research and forensic applications
Catalog No. 50-291-9372 Supplier Cayman Chemical Supplier No. 3723810 MG
An analytical reference standard categorized as a diuretic; has been found as a potential doping agent in urine samples from athletes but can be formed from proguanil; intended for research and forensic applications