Learn More
Ethos Biosciences Chlorazol Black E 2oz
Catalog No. 5000819194
Encompass_Preferred
Chlorazole Black E is a dark black to brownish-black solution with strong affinity for cellulose and chitin This solution is widely used as a biological stain for plant histology differentiating cellulose from lignin and staining chitain in fungal cell walls Also serves as a textile dye for cotton rayon leather and paper and as a pH indicator
Catalog No. 50-008-19194 Supplier Ethos Biosciences Supplier No. 695002D
Chlorazole Black E is a dark black to brownish-black solution with strong affinity for cellulose and chitin This solution is widely used as a biological stain for plant histology differentiating cellulose from lignin and staining chitain in fungal cell walls Also serves as a textile dye for cotton rayon leather and paper and as a pH indicator