Learn More
TARGETMOL CHEMICALS INC CHLORCYCLIZINE 50MG
SDP
Catalog No. 502578969
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CHLORCYCLIZINE is a histamine H1 antagonist and a potent hepatitis C virus (HCV) entry inhibitor. purity: 98%
Catalog No. 50-257-8969 Supplier TARGETMOL CHEMICALS INC Supplier No. T971750MG
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CHLORCYCLIZINE is a histamine H1 antagonist and a potent hepatitis C virus (HCV) entry inhibitor. purity: 98%