Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorcyclizine HCl, MP Biomedicals

Catalog No. ICN201858 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201858 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN201858 Supplier MP Biomedicals, Inc Supplier No. 0520185810
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14362-31-3
Molecular Formula C18H22Cl2N2
Molecular Weight (g/mol) 337.29
MDL Number MFCD00035329
InChI Key MSIJLVMSKDXAQN-UHFFFAOYNA-N
Synonym chlorcyclizine hydrochloride, chlorcyclizine hcl, histantin, eramide, perazil, di-paralene, chlorcyclizinium chloride, chlorcyclizine chloride, chlorcyclizine monohydrochloride, 1-p-chlorobenzhydryl-4-methylpiperazine monohydrochloride
PubChem CID 62413
IUPAC Name hydrogen 1-[(4-chlorophenyl)(phenyl)methyl]-4-methylpiperazine chloride
SMILES [H+].[Cl-].CN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1

Specifications

Quantity 10 g
Formula Weight 300.85
Chemical Name or Material Chlorcyclizine HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.