Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chlorfenazole (o-chlorobenzimidazole) | 3574-96-7 | MFCD00051632 | 99.9% | 228.68 g/mol | C13H9ClN2 | 250 MG
SDP

Catalog No. 5000209692
Encompass_Preferred

Chlorfenazole (o-chlorobenzimidazole) is a benzimidazole-based compound primarily used in research of intestinal nematode diseases.

  • Used in research of intestinal nematode diseases.
  • Cas number 3574-96-7.
  • Chemical formula C13H9ClN2.
  • Molecular weight 228.68 g/mol.
  • Purity 99.9% (HPLC) as shown on manufacturer certificate of analysis.
  • Target: parasite; pathway: anti-infection.
  • Recommended storage: room temperature (3 years); in solvent -80°C (2 years) or -20°C (1 year).

Catalog No. 50-002-09692 Supplier Medchemexpress LLC Supplier No. 250MGHYW087946
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorfenazole (o-chlorobenzimidazole) is a benzimidazole-based compound primarily used in research of intestinal nematode diseases.

  • Used in research of intestinal nematode diseases.
  • Cas number 3574-96-7.
  • Chemical formula C13H9ClN2.
  • Molecular weight 228.68 g/mol.
  • Purity 99.9% (HPLC) as shown on manufacturer certificate of analysis.
  • Target: parasite; pathway: anti-infection.
  • Recommended storage: room temperature (3 years); in solvent -80°C (2 years) or -20°C (1 year).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.