Learn More
AdipoGen Chlorferone
SDP
Supplier:  AdipoGen CDXC0351G005
Encompass_Preferred
Chemical. CAS 6174-86-3. Formula C10H7ClO3. MW 210.61. Synthetic. Major metabolite and hydrolysis product of coumaphos, an organophosphate pesticide. Shown to have acetylcholinesterase and butyrylcholinesterase inhibitor activity and moderate to weak alpha-glucosidase inhibitory and antioxidant scavenging activity. Also used as a building block in the synthesis of fluorogenic substrates for the detection of bacterial beta-Galactosidase.
Catalog No. 50-256-1369