Learn More
Selleck Chemical LLC Chlorhexidine S5397-25mg
SDP
Catalog No. 503111524
Encompass_Preferred
Chlorhexidine is a disinfectant and topical anti-infective agent that is active against Gram-positive and Gram-negative organisms facultative anaerobes aerobes and yeasts
Catalog No. 50-311-1524 Supplier Selleck Chemical LLC Supplier No. S539725MG
Chlorhexidine is a disinfectant and topical anti-infective agent that is active against Gram-positive and Gram-negative organisms facultative anaerobes aerobes and yeasts