Learn More
Apexbio Technology LLC CHLORHEXIDINE-DIACETATE-200MG
SDP
Catalog No. 5000569583
Encompass_Preferred
Chlorhexidine diacetate (CAS No 56-95-1) is a widely researched antimicrobial agent in the biomedical field known for its effectiveness against a broad spectrum of microorganisms Originating from its development as a potent disinfectant its mechanism of action involves disrupting bacterial cell membranes leading to cell death Its in vitro activity is often observed in the nanomolar to low micromolar range demonstrating strong efficacy at relatively low concentrations Chlorhexidine diacetate is commonly utilized in various experimental applications including cell culture studies animal model testing and drug screening assays In these contexts it serves as a benchmark for evaluating antimicrobial effectiveness or as a control compound in comparative studies Experimental concentrations typically depend on the specific research design Its widespread use underscores its importance in understanding microbial resistance and developing new antimicrobial strategies
Catalog No. 50-005-69583 Supplier Apexbio Technology LLC Supplier No. C7112200
Chlorhexidine diacetate (CAS No 56-95-1) is a widely researched antimicrobial agent in the biomedical field known for its effectiveness against a broad spectrum of microorganisms Originating from its development as a potent disinfectant its mechanism of action involves disrupting bacterial cell membranes leading to cell death Its in vitro activity is often observed in the nanomolar to low micromolar range demonstrating strong efficacy at relatively low concentrations Chlorhexidine diacetate is commonly utilized in various experimental applications including cell culture studies animal model testing and drug screening assays In these contexts it serves as a benchmark for evaluating antimicrobial effectiveness or as a control compound in comparative studies Experimental concentrations typically depend on the specific research design Its widespread use underscores its importance in understanding microbial resistance and developing new antimicrobial strategies