Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine digluconate, 20% w/v aq. soln., non-sterile

Catalog No. AA41385AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
4 x 500 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41385AC 25 mL
AA41385AE 100 mL
AA41385AP 500 mL
AA41385AY 4 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA41385AC Supplier Thermo Scientific Chemicals Supplier No. 041385AC
Only null left
Add to Cart

CAS: 18472-51-0 | C34H54Cl2N10O14 | 897.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Chlorhexidine digluconate, 20% w/v aq. Soln is used as an oral antiseptic agent and anti-microbial agent. It is an active ingredient in germicidal mouthwash, pet shampoo, surgical lubricants and wipes and hand soap. It is utilized in the study to know the role of essential oils for improving skin antisepsis. It plays a vital role as disinfectants and preservative in pharmaceutical products. It is also used as an additive in cosmetics such as creams, toothpaste and deodorants.

Solubility
Miscible with water, ethanol and acetone.

Notes
Store in a cool place. Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18472-51-0
Molecular Formula C34H54Cl2N10O14
Molecular Weight (g/mol) 897.76
MDL Number MFCD00083599
InChI Key YZIYKJHYYHPJIB-UUPCJSQJSA-N
Synonym peridex, periogard, chlorhexidine gluconate, 1,1'-hexamethylene bis 5-p-chlorophenyl biguanide di-d-gluconate
PubChem CID 60148207
IUPAC Name (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;sulfane
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

Color Colorless
Density 1.06 g/mL
Assay Percent Range 20% w/v aq. soln.
Quantity 25 mL
Sensitivity Light sensitive
Formula Weight 897.76
Concentration or Composition (by Analyte or Components) 20% w/v aq. soln.
Physical Form Liquid
Chemical Name or Material Chlorhexidine digluconate, Non-sterile
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 242-354-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.