Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine Digluconate Aqueous Solution 0.2 MP Biomedicals

Catalog No. ICN19136025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19136025 25 mL
ICN19136090 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19136025 Supplier MP Biomedicals, Inc Supplier No. 0219136025
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18472-51-0
Molecular Formula C34H54Cl2N10O14
Molecular Weight (g/mol) 897.76
MDL Number MFCD00083599
InChI Key YZIYKJHYYHPJIB-UUPCJSQJSA-N
Synonym peridex, periogard, chlorhexidine gluconate, 1,1'-hexamethylene bis 5-p-chlorophenyl biguanide di-d-gluconate
PubChem CID 60148207
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(O)=O.NC(NC1=CC=C(Cl)C=C1)=NC(N)=NCCCCCCN=C(N)N=C(N)NC1=CC=C(Cl)C=C1

Specifications

Quantity 25 mL
IUPAC Name bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid); N'-(6-{[amino({amino[(4-chlorophenyl)amino]methylidene}amino)methylidene]amino}hexyl)-1-({amino[(4-chlorophenyl)amino]methylidene}amino)methanimidamide
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.