Learn More
Bioworld Chlorhexidine DiGluconate, 100 mL
Supplier:  Bioworld 403000761
Encompass_Preferred
Chlorhexidine is a bisguanide antiseptic and disinfectant, which is bactericidal or bacteriostatic against a wide range of gram-positive and gram-negative bacteria. It inhibits mycobacterium, fungi and some viruses. Chlorhexidine is most active at a neutral or slightly acidic pH. It is used to disinfect skin, to clean instruments and hard surfaces at a concentration of 0.05% to 0.5% in water. Technical Information:20% aqueous solutionApplication: Antiseptic, disinfectant agent, antimicrobialSubstance is listed in Toxic Substance Control Act (TSCA) inventoryAppearance: LiquidDensity : 1.06 g/mlMelting Point: 134 °C
Catalog No. 50-255-678