Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine Free Base, >85%, MP Biomedicals™

Catalog No. ICN19135805 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19135805 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19135805 Supplier MP Biomedicals, Inc Supplier No. 0219135805
Add to Cart
Add to Cart
Molecular biology; Cell biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-56-1
Molecular Formula C22H30Cl2N10
Molecular Weight (g/mol) 505.452
MDL Number MFCD00009673
InChI Key GHXZTYHSJHQHIJ-UHFFFAOYSA-N
Synonym chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
PubChem CID 9552079
ChEBI CHEBI:3614
IUPAC Name (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
SMILES C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl

Specifications

Color White
Quantity 5 g
Physical Form Solid
Assay Percent Range >85%
Formula Weight 505.5
Chemical Name or Material Chlorhexidine Free Base, Free Base
EINECSNumber 200-238-7
Recommended Storage Store at +4 °C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.