Learn More
Apexbio Technology LLC Chlorhexidine HCl 3697-42-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092638
Encompass_Preferred
Chlorhexidine HCl (CAS 3697-42-5) is an antiseptic agent designed to interact with microbial cell membranes thereby increasing membrane permeability and disrupting membrane integrity Chlorhexidine HCl exerts its biological activity primarily through binding to negatively charged surfaces of bacterial membranes resulting in leakage of cellular contents and consequent microbial cell death In microbiological assays chlorhexidine HCl demonstrates broad-spectrum antimicrobial activity with reported IC50 values against various microbial strains generally in the micromolar concentration range though specific values depend on the target organism and experimental conditions Based on these pharmacological properties chlorhexidine HCl holds research potential in antimicrobial susceptibility testing and as a control compound in comparative studies of novel antimicrobial agents
Catalog No. 50-000-92638 Supplier Apexbio Technology LLC Supplier No. B16955.1
Chlorhexidine HCl (CAS 3697-42-5) is an antiseptic agent designed to interact with microbial cell membranes thereby increasing membrane permeability and disrupting membrane integrity Chlorhexidine HCl exerts its biological activity primarily through binding to negatively charged surfaces of bacterial membranes resulting in leakage of cellular contents and consequent microbial cell death In microbiological assays chlorhexidine HCl demonstrates broad-spectrum antimicrobial activity with reported IC50 values against various microbial strains generally in the micromolar concentration range though specific values depend on the target organism and experimental conditions Based on these pharmacological properties chlorhexidine HCl holds research potential in antimicrobial susceptibility testing and as a control compound in comparative studies of novel antimicrobial agents