Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorhexidine HCl, G-Biosciences
SCI_ED

Catalog No. S930074
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S930074 5 g
S000014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S930074 Supplier G-Biosciences Supplier No. RC966
Add to Cart
Add to Cart

Chlorhexidine HCl is the cationic salt form of chlorhexidine, a bactericidal detergent.

Chlorhexidine HCl has strong bactericidal activity and is used for in vitro bactericidal assays as a positive control.

Chemical Identifiers

Molecular Weight (g/mol) 578.37 g/mol

Specifications

Boiling Point 699.3°C
Quantity 5 g
Percent Purity >95%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.