Learn More
ERA WATERS CORP CHLORIDE - 1000 MG/L, 125 ML
SDP
Catalog No. NC0350738
Encompass
Chloride - 1000 mg/l, 125 ml One 125 mL standard containing Chloride at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.
Catalog No. NC0350738 Supplier ERA WATERS CORP Supplier No. 047
Chloride - 1000 mg/l, 125 ml One 125 mL standard containing Chloride at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.