Learn More
SPEX CERTIPREP LLC 3P Chloride from Sodium Chloride at 10,000 ug/mL in H2O, 250 mL
Catalog No. NC1815677
Encompass_Preferred
(HPS by Zeptometrix) Chloride Anion IC Single-Component Standard from Sodium Chloride at 10,000 ug/mL in H2O.
Catalog No. NC1815677 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICCL10M250
(HPS by Zeptometrix) Chloride Anion IC Single-Component Standard from Sodium Chloride at 10,000 ug/mL in H2O.