Learn More
Hach Company Chloride Standard Solution, 12,500 mg/L as Cl- (NIST), pk/16 - 10 mL Voluette Ampules
Catalog No. 501994970 Shop All Hach Company Products
Encompass
For accuracy checks in chloride determinations
Catalog No. 50-199-4970 Supplier Hach Company Supplier No. 1425010
For accuracy checks in chloride determinations