Learn More
Hach Company Chloride 2 Indicator Powder Pillows, 100 mL, pk/50
Catalog No. 501994508 Shop All Hach Company Products
Encompass
Indicator used in the determination of chloride by argentometric titration with silver nitrate. Sample size: 100mL. Pack of 50 Powder Pillows
Catalog No. 50-199-4508 Supplier Hach Company Supplier No. 105766
Indicator used in the determination of chloride by argentometric titration with silver nitrate. Sample size: 100mL. Pack of 50 Powder Pillows