Learn More
SPEX CERTIPREP LLC 3P CHLORIDE 250 ML 1000PPM
Catalog No. NC9636381
Encompass_Preferred
(HPS by Zeptometrix) 1000 ug/mL Chloride in H2O; Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9636381 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICCLM250
(HPS by Zeptometrix) 1000 ug/mL Chloride in H2O; Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.