Learn More
Hach Company Chloride Ionic Strength Adjustor (ISA) Powder Pillows, pk/100
Catalog No. 501995785 Shop All Hach Company Products
Encompass
Ionic Strength Adjustor (ISA) for Chloride determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Chloride level readings. Eliminate most interferences. Pack of 100 powder pillows.
Catalog No. 50-199-5785 Supplier Hach Company Supplier No. 2318069
Ionic Strength Adjustor (ISA) for Chloride determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Chloride level readings. Eliminate most interferences. Pack of 100 powder pillows.