Learn More
Hach Company Intellical™ ISECL181 Chloride (Cl⁻) Ion Selective Electrode (ISE), 3 m cable
Catalog No. 501997719 Shop All Hach Company Products
Encompass
Intellical™ ISECL181 is a digital, combination, Chloride ion selective electrode (ISE) with a with a non-refillable gel driTEK reference with a double junction, ceramic porous pin and porous PTFE annular ring, and built-in temperature sensor. The electrode measures Chloride concentration in water samples. The laboratory version of this ISE is shockproff with its Epoxy plastic body.The Intellical ISECL181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISECL181’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE. The ISECL181 is ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications.
Catalog No. 50-199-7719 Supplier Hach Company Supplier No. ISECL18103
Intellical™ ISECL181 is a digital, combination, Chloride ion selective electrode (ISE) with a with a non-refillable gel driTEK reference with a double junction, ceramic porous pin and porous PTFE annular ring, and built-in temperature sensor. The electrode measures Chloride concentration in water samples. The laboratory version of this ISE is shockproff with its Epoxy plastic body.The Intellical ISECL181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISECL181’s solid-state sensor design eliminates membrane replacement and allows for dry storage of the ISE. The ISECL181 is ideal for measuring chloride concentrations in wastewater, drinking water and general water quality applications.