Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloride Standard, 0.100 Normal 100 mEq/L NaCl, Reagents

Catalog No. 01335641
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-641 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-641 Supplier Reagents Holdings Llc Supplier No. CC155000500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chloride Standard, 0.100 Normal 100 mEq/L NaCl, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Chloride Standard, 0.100 Normal 100 mEq/L NaCl
Packaging Poly Bottle
CAS 7732-18-5, 7647-14-5
Specific Gravity 1.002
Physical Form Liquid
Molecular Formula NaCl/H2O
Color Colorless
pH 6.7 to 7.3
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Water Analysis
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.