Learn More
Aqua Solutions Chloride Standard 10,000 ppm Cl 1 ml = 10 mg Cl, NIST Traceable (100ML)
SDP
Catalog No. NC0325798
Encompass
Chloride Standard 10,000 ppm Cl 1 ml = 10 mg Cl, NIST Traceable (100ML)
Catalog No. NC0325798 Supplier Aqua Solutions Supplier No. AM414
Chloride Standard 10,000 ppm Cl 1 ml = 10 mg Cl, NIST Traceable (100ML)